Assay Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177606.4 | NP_808274.2 | 13 - 14 | 2286 | 91 | |
| GenBank mRNA | AK029252.1 | - | 13 - 14 | 2288 | 91 | |
| AK173337.1 | - | 10 - 11 | 1941 | 91 | ||
| BC125583.1 | - | 13 - 14 | 2251 | 40130098 | 91 | |
| BC137804.1 | - | 13 - 14 | 2248 | 8860823 | 91 | |
More Information
Set Membership: |
|

Plekhh2
Genomic Map