Assay Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177606.4 | NP_808274.2 | 26 - 27 | 4019 | 68 | |
| GenBank mRNA | AK029252.1 | - | 26 - 27 | 4021 | 68 | |
| AK086794.1 | - | 6 - 7 | 849 | 68 | ||
| AK173337.1 | - | 23 - 24 | 3674 | 68 | ||
| BC125583.1 | - | 26 - 27 | 3984 | 40130098 | 68 | |
| BC137804.1 | - | 26 - 27 | 3981 | 8860823 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plekhh2
Genomic Map