Assay Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177608.3 | NP_808276.2 | 1 - 2 | 274 | 98 | |
| GenBank mRNA | AK049924.1 | - | 1 - 2 | 275 | 98 | |
| AK145135.1 | - | 1 - 2 | 159 | 98 | ||
| AK169713.1 | - | 1 - 2 | 161 | 98 | ||
| AK172920.1 | - | 1 - 2 | 143 | 98 | ||
| BC095939.1 | - | 1 - 2 | 141 | 98 | ||
| BC141056.1 | - | 1 - 2 | 234 | 9055668 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049924.1, AK169713.1, BC095939.1, BC141056.1, NM_177608.3 transcripts.
Set Membership: |
|

Secisbp2l
Genomic Map