Assay Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177608.3 | NP_808276.2 | 3 - 4 | 779 | 92 | |
| GenBank mRNA | AK049924.1 | - | 3 - 4 | 780 | 92 | |
| AK169713.1 | - | 3 - 4 | 666 | 92 | ||
| AK172920.1 | - | 3 - 4 | 648 | 92 | ||
| BC095939.1 | - | 3 - 4 | 646 | 92 | ||
| BC141056.1 | - | 3 - 4 | 739 | 9055668 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Secisbp2l
Genomic Map