Assay Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177608.3 | NP_808276.2 | 10 - 11 | 1662 | 135 | |
| GenBank mRNA | AK049924.1 | - | 10 - 11 | 1663 | 137 | |
| AK145135.1 | - | 10 - 11 | 1548 | 135 | ||
| AK169713.1 | - | 10 - 11 | 1549 | 135 | ||
| AK172920.1 | - | 10 - 11 | 1531 | 133 | ||
| BC071239.1 | - | 3 - 4 | 290 | 135 | ||
| BC141056.1 | - | 10 - 11 | 1622 | 9055668 | 135 | |
| CJ198024.1 | - | 1 - 2 | 164 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Secisbp2l
Genomic Map