Assay Details
| Gene Symbol: Ccdc157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164620.1 | NP_001158092.1 | 2 - 3 | 288 | 64 | |
| NM_177616.3 | NP_808284.2 | 2 - 3 | 288 | 64 | ||
| GenBank mRNA | AK076934.1 | - | 2 - 3 | 216 | 64 | |
| AK133165.1 | - | 2 - 3 | 289 | 64 | ||
| AK220409.1 | - | 2 - 3 | 269 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076934.1, NM_001164620.1, NM_177616.3 transcripts.
Set Membership: |
|

Ccdc157
Genomic Map