Assay Details
| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161365.1 | NP_001154837.1 | 5 - 6 | 633 | 77 | |
| NM_177620.4 | NP_808288.2 | 5 - 6 | 744 | 77 | ||
| GenBank mRNA | AK032205.1 | - | 5 - 6 | 745 | 77 | |
| AK154522.1 | - | 5 - 6 | 548 | 77 | ||
| AK154963.1 | - | 5 - 6 | 634 | 77 | ||
| AK166196.1 | - | 5 - 6 | 590 | 77 | ||
| AK171301.1 | - | 5 - 6 | 557 | 77 | ||
| AK202814.1 | - | 3 - 4 | 264 | 77 | ||
| AY265356.1 | - | 5 - 6 | 730 | 77 | ||
| BC137907.1 | - | 5 - 6 | 672 | 8860926 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rin3
Genomic Map