Assay Details
| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122738.1 | NP_001116210.1 | 5 - 6 | 1705 | 85 | |
| NM_177645.4 | NP_808313.3 | 5 - 6 | 1810 | 85 | ||
| GenBank mRNA | AK082897.1 | - | 4 - 5 | 525 | 85 | |
| AK145440.1 | - | 5 - 6 | 1782 | 85 | ||
| AK220534.1 | - | 5 - 6 | 1761 | 85 | ||
| BC141036.1 | - | 6 - 7 | 1720 | 9055648 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kansl1l
Genomic Map