Assay Details
| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122738.1 | NP_001116210.1 | 8 - 9 | 2172 | 137 | |
| GenBank mRNA | AK082897.1 | - | 7 - 8 | 992 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kansl1l
Genomic Map