Assay Details
| Gene Symbol: Rassf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178045.4 | NP_835146.3 | 3 - 4 | 279 | 70 | |
| GenBank mRNA | AK036528.1 | - | 3 - 4 | 243 | 70 | |
| AK053578.1 | - | 3 - 4 | 241 | 70 | ||
| AK089460.1 | - | 3 - 4 | 236 | 70 | ||
| AK139284.1 | - | 3 - 4 | 226 | 70 | ||
| AK151561.1 | - | 3 - 4 | 243 | 70 | ||
| AK152639.1 | - | 3 - 4 | 243 | 70 | ||
| AK160584.1 | - | 3 - 4 | 253 | 70 | ||
| AK164357.1 | - | 3 - 4 | 242 | 70 | ||
| BC023245.1 | - | 3 - 4 | 232 | 4459337 | 70 | |
| BC060709.1 | - | 3 - 4 | 234 | 6409692 | 70 | |
| BY170346.1 | - | 3 - 4 | 280 | 70 | ||
More Information
Set Membership: |
|

Rassf4
Genomic Map