Assay Details
| Gene Symbol: Atl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286647.1 | NP_001273576.1 | 9 - 10 | 978 | 69 | |
| NM_019717.3 | NP_062691.3 | 10 - 11 | 1223 | 69 | ||
| NM_178050.4 | NP_835151.2 | 9 - 10 | 978 | 69 | ||
| GenBank mRNA | AK032050.1 | - | 1 - 2 | 1574 | 69 | |
| AK038697.1 | - | 9 - 10 | 906 | 69 | ||
| AK135988.1 | - | 9 - 10 | 869 | 69 | ||
| BC006934.1 | - | 2 - 3 | 106 | 69 | ||
| BC051095.1 | - | 1 - 2 | 47 | 69 | ||
| BC055466.1 | - | 7 - 8 | 581 | 69 | ||
| BC060501.1 | - | 10 - 11 | 1128 | 5362692 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032050.1 transcripts.
Set Membership: |
|

Atl2
Genomic Map