Assay Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177231.2 | NP_796205.1 | 10 - 11 | 1069 | 69 | |
| NM_178220.3 | NP_835738.1 | 10 - 11 | 1069 | 69 | ||
| GenBank mRNA | AK052588.1 | - | 10 - 11 | 846 | 69 | |
| AK090090.1 | - | 10 - 11 | 896 | 69 | ||
| AK144054.1 | - | 10 - 11 | 868 | 69 | ||
| AK147508.1 | - | 10 - 11 | 1070 | 69 | ||
| AK147566.1 | - | 10 - 11 | 1091 | 69 | ||
| AK165514.1 | - | 10 - 11 | 868 | 69 | ||
| BC108969.1 | - | 10 - 11 | 836 | 40047224 | 69 | |
| BC108970.1 | - | 10 - 11 | 836 | 40047225 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arrb1
Genomic Map