Assay Details
| Gene Symbol: Reps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290633.1 | NP_001277562.1 | 7 - 8 | 1079 | 67 | |
| NM_178256.4 | NP_839987.2 | 7 - 8 | 1082 | 67 | ||
| GenBank mRNA | AK147418.1 | - | 7 - 8 | 730 | 67 | |
| AK147432.1 | - | 4 - 5 | 361 | 67 | ||
| AY263368.1 | - | 7 - 8 | 1027 | 67 | ||
| BC139012.1 | - | 7 - 8 | 958 | 8862034 | 67 | |
| BC145500.1 | - | 7 - 8 | 955 | 9054063 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reps2
Genomic Map