Assay Details
| Gene Symbol: Cdc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178347.4 | NP_848124.1 | 8 - 9 | 961 | 72 | |
| GenBank mRNA | AK031459.1 | - | 8 - 9 | 944 | 72 | |
| AK038523.1 | - | 8 - 9 | 944 | 72 | ||
| AK047789.1 | - | 8 - 9 | 944 | 72 | ||
| AK048246.1 | - | 8 - 9 | 944 | 72 | ||
| AK052567.1 | - | 8 - 9 | 961 | 72 | ||
| AK075948.1 | - | 8 - 9 | 962 | 72 | ||
| AK081461.1 | - | 8 - 9 | 962 | 72 | ||
| AK169677.1 | - | 4 - 5 | 435 | 72 | ||
| BC059013.1 | - | 8 - 9 | 930 | 6813730 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc23
Genomic Map