Assay Details
| Gene Symbol: Prss33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081399.2 | NP_001074868.2 | 5 - 6 | 719 | 71 | |
| GenBank mRNA | AY262280.1 | - | 5 - 6 | 695 | 71 | |
| BC101950.1 | - | 5 - 6 | 695 | 40044230 | 71 | |
| BC103541.1 | - | 6 - 7 | 791 | 40044224 | 71 | |
| BC103542.1 | - | 5 - 6 | 695 | 40044219 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | NM_001081399.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | AY262280.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | BC101950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | BC103541.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | BC103542.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | NM_001081399.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | AY262280.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | BC101950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | BC103541.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342654 | BC103542.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125627 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150011 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | NM_001081399.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | AY262280.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | BC101950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | BC103541.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | BC103542.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | NM_001081399.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | AY262280.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | BC101950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | BC103541.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342654 | BC103542.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150011 | - | - |
| Whole Mouse Genome G4122A | A_52_P342654 | NM_001081399.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | AY262280.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | BC101950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | BC103541.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | BC103542.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | NM_001081399.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | AY262280.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | BC101950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | BC103541.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P342654 | BC103542.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150011 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127270 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127244 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127236 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127237 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31093989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13127228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13127258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31093943 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10448387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448384 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10448383 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700014078.mm.1 | NM_001081399.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014078.mm.1 | NM_001081399.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014078.mm.1 | TC1700001607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014079.mm.1 | NM_001081399.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014079.mm.1 | NM_001081399.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014079.mm.1 | TC1700001607.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 428610 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1500450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231590 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231590 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231590 | - | - |

Prss33
Genomic Map