Assay Details
| Gene Symbol: Snupn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178374.3 | NP_848461.1 | 3 - 4 | 347 | 75 | |
| GenBank mRNA | AK002717.1 | - | 3 - 4 | 363 | 75 | |
| AK013151.1 | - | 3 - 4 | 357 | 75 | ||
| AK155315.1 | - | 3 - 4 | 364 | 75 | ||
| BC023317.1 | - | 327 | 75 | |||
| BC049638.1 | - | 3 - 4 | 347 | 6589249 | 75 | |
| BC061258.1 | - | 3 - 4 | 352 | 30251661 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snupn
Genomic Map