Assay Details
| Gene Symbol: Iqcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178378.3 | NP_848465.1 | 7 - 8 | 921 | 72 | |
| GenBank mRNA | AK010269.1 | - | 4 - 5 | 470 | 72 | |
| AK146422.1 | - | 5 - 6 | 659 | 72 | ||
| BC049990.1 | - | 7 - 8 | 905 | 6310844 | 72 | |
More Information
Set Membership: |
|

Iqcg
Genomic Map