| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500004130.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004130.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004130.mm.1 |
NM_178394.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004131.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004131.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004131.mm.1 |
NM_178394.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004125.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004125.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004125.mm.1 |
NM_178394.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004127.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004127.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004127.mm.1 |
NM_178394.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004128.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004128.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004128.mm.1 |
NM_178394.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004134.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004134.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004134.mm.1 |
NM_178394.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004126.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004126.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004126.mm.1 |
NM_178394.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004135.mm.1 |
TC0500000418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004135.mm.1 |
NM_178394.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004135.mm.1 |
NM_178394.4 |
Refseq |