Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500018746.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018746.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018746.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018746.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018747.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018747.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018747.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018747.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018725.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018725.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018725.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018725.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018729.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018729.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018729.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018729.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018744.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018744.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018744.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018744.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018745.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018745.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018745.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018745.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018748.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018748.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018750.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018750.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018750.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018738.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018738.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018738.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018738.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018726.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018726.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018726.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018726.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018742.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018742.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018742.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018742.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018733.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018733.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018733.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018733.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018735.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018735.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018735.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018735.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018736.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018736.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018736.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018736.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018753.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018753.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018753.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018732.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018732.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018732.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018732.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018737.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018737.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018737.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018737.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018741.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018741.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018741.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018741.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018743.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018743.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018743.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018743.mm.1 |
NM_178403.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018749.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018749.mm.1 |
NM_001289781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018751.mm.1 |
TC0500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018751.mm.1 |
NM_001289780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018751.mm.1 |
NM_178403.5 |
Refseq |