Assay Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178405.3 | NP_848492.1 | 12 - 13 | 1823 | 68 | |
| GenBank mRNA | AK052863.1 | - | 12 - 13 | 1760 | 68 | |
| AK129212.1 | - | 12 - 13 | 1736 | 68 | ||
| AK137602.1 | - | 12 - 13 | 1759 | 68 | ||
| AK147331.1 | - | 12 - 13 | 1755 | 68 | ||
| AK148775.1 | - | 12 - 13 | 1755 | 68 | ||
| AK161840.1 | - | 12 - 13 | 1755 | 68 | ||
| AK164509.1 | - | 12 - 13 | 1759 | 68 | ||
| AK168703.1 | - | 12 - 13 | 1755 | 68 | ||
| BC025807.1 | - | 1 - 2 | 51 | 68 | ||
| BC036127.1 | - | 12 - 13 | 1747 | 4955003 | 68 | |
| BC041774.1 | - | 9 - 10 | 1473 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a2
Genomic Map