Assay Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178405.3 | NP_848492.1 | 17 - 18 | 2614 | 73 | |
| GenBank mRNA | AK129212.1 | - | 17 - 18 | 2527 | 73 | |
| AK137602.1 | - | 17 - 18 | 2550 | 73 | ||
| AK147331.1 | - | 17 - 18 | 2546 | 73 | ||
| AK148775.1 | - | 17 - 18 | 2546 | 73 | ||
| AK161840.1 | - | 17 - 18 | 2546 | 73 | ||
| AK164509.1 | - | 17 - 18 | 2550 | 73 | ||
| AK168703.1 | - | 17 - 18 | 2546 | 73 | ||
| BC025807.1 | - | 6 - 7 | 842 | 73 | ||
| BC036127.1 | - | 17 - 18 | 4955003 | 73 | ||
| BC041774.1 | - | 14 - 15 | 2264 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a2
Genomic Map