Assay Details
| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164564.1 | NP_001158036.1 | 5 - 6 | 506 | 60 | |
| NM_178444.4 | NP_848538.3 | 6 - 7 | 731 | 60 | ||
| NM_198724.2 | NP_942017.2 | 6 - 7 | 730 | 60 | ||
| NM_198725.2 | NP_942018.2 | 6 - 7 | 730 | 60 | ||
| GenBank mRNA | AF184973.1 | - | 6 - 7 | 681 | 60 | |
| AK002601.1 | - | 6 - 7 | 726 | 60 | ||
| AK139377.1 | - | 5 - 6 | 501 | 60 | ||
| AK156158.1 | - | 6 - 7 | 732 | 60 | ||
| AY239289.1 | - | 6 - 7 | 692 | 60 | ||
| AY239290.1 | - | 6 - 7 | 714 | 60 | ||
| AY309459.1 | - | 6 - 7 | 704 | 60 | ||
| BC024610.1 | - | 6 - 7 | 714 | 4159675 | 60 | |
More Information
Set Membership: |
|

Egfl7
Genomic Map