Assay Details
| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164564.1 | NP_001158036.1 | 6 - 7 | 669 | 64 | |
| NM_178444.4 | NP_848538.3 | 7 - 8 | 894 | 64 | ||
| NM_198724.2 | NP_942017.2 | 7 - 8 | 893 | 64 | ||
| NM_198725.2 | NP_942018.2 | 7 - 8 | 893 | 64 | ||
| GenBank mRNA | AF184973.1 | - | 7 - 8 | 844 | 64 | |
| AK002601.1 | - | 7 - 8 | 889 | 64 | ||
| AK139377.1 | - | 6 - 7 | 664 | 64 | ||
| AK156158.1 | - | 7 - 8 | 896 | 64 | ||
| AY239289.1 | - | 7 - 8 | 855 | 64 | ||
| AY239290.1 | - | 7 - 8 | 877 | 64 | ||
| AY309459.1 | - | 7 - 8 | 867 | 64 | ||
| BC024610.1 | - | 7 - 8 | 877 | 4159675 | 64 | |
More Information
Set Membership: |
|

Egfl7
Genomic Map