Assay Details
| Gene Symbol: Abhd16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178592.3 | NP_848707.1 | 13 - 14 | 1221 | 83 | |
| GenBank mRNA | AK195312.1 | - | 1 - 2 | 103 | 83 | |
| BC029114.1 | - | 13 - 14 | 1221 | 2648618 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Abhd16a
Genomic Map