Assay Details
| Gene Symbol: Abhd16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178592.3 | NP_848707.1 | 5 - 6 | 459 | 71 | |
| GenBank mRNA | BC029114.1 | - | 5 - 6 | 459 | 2648618 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd16a
Genomic Map