Assay Details
| Gene Symbol: Ptrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178595.3 | NP_848710.1 | 2 - 3 | 295 | 78 | |
| GenBank mRNA | AK075833.1 | - | 2 - 3 | 296 | 78 | |
| AK132137.1 | - | 331 | 78 | |||
| BC098218.1 | - | 2 - 3 | 335 | 3466952 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptrh1
Genomic Map