Assay Details
| Gene Symbol: Fbxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178624.6 | NP_848739.1 | 8 - 9 | 637 | 121 | |
| GenBank mRNA | AK013142.1 | - | 9 - 10 | 771 | 121 | |
| AK039010.1 | - | 8 - 9 | 637 | 121 | ||
| AK044693.1 | - | 8 - 9 | 634 | 121 | ||
| AK045742.1 | - | 8 - 9 | 624 | 121 | ||
| AK089994.1 | - | 8 - 9 | 613 | 121 | ||
| AK141177.1 | - | 8 - 9 | 627 | 121 | ||
| AK163472.1 | - | 8 - 9 | 633 | 121 | ||
| BC051530.1 | - | 4 - 5 | 451 | 121 | ||
| BC096582.1 | - | 8 - 9 | 623 | 3963599 | 121 | |
| BC145665.1 | - | 8 - 9 | 617 | 40130968 | 121 | |
| BC145998.1 | - | 8 - 9 | 617 | 40131301 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl2
Genomic Map