Assay Details
| Gene Symbol: Fbxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178624.6 | NP_848739.1 | 10 - 11 | 843 | 73 | |
| GenBank mRNA | AK013142.1 | - | 11 - 12 | 977 | 73 | |
| AK039010.1 | - | 10 - 11 | 843 | 73 | ||
| AK045742.1 | - | 10 - 11 | 830 | 73 | ||
| AK089994.1 | - | 10 - 11 | 819 | 73 | ||
| AK141177.1 | - | 10 - 11 | 833 | 73 | ||
| AK163472.1 | - | 10 - 11 | 839 | 73 | ||
| BC051530.1 | - | 6 - 7 | 657 | 73 | ||
| BC096582.1 | - | 10 - 11 | 829 | 3963599 | 73 | |
| BC145665.1 | - | 10 - 11 | 823 | 40130968 | 73 | |
| BC145998.1 | - | 10 - 11 | 823 | 40131301 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl2
Genomic Map