Assay Details
| Gene Symbol: Sfxn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178639.4 | NP_848754.1 | 12 - 13 | 835 | 58 | |
| GenBank mRNA | AK036285.1 | - | 12 - 13 | 833 | 58 | |
| AK082073.1 | - | 12 - 13 | 833 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sfxn5
Genomic Map