Assay Details
| Gene Symbol: Sfxn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178639.4 | NP_848754.1 | 6 - 7 | 370 | 75 | |
| GenBank mRNA | AF325264.1 | - | 6 - 7 | 388 | 75 | |
| AK036285.1 | - | 6 - 7 | 368 | 75 | ||
| AK043706.1 | - | 6 - 7 | 369 | 75 | ||
| AK082073.1 | - | 6 - 7 | 368 | 75 | ||
| BB628882.1 | - | 6 - 7 | 371 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sfxn5
Genomic Map