Assay Details
| Gene Symbol: Tigd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178646.4 | NP_848761.2 | 1 | 1379 | 66 | |
| GenBank mRNA | AK036241.1 | - | 1 | 1352 | 66 | |
| AK051177.1 | - | 2 | 1369 | 66 | ||
| AK086663.1 | - | 1 | 1356 | 66 | ||
| AK165093.1 | - | 1 | 1357 | 66 | ||
| BC099838.1 | - | 1 | 1379 | 5703010 | 66 | |
| BC138289.1 | - | 1 | 1353 | 8861308 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344888 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344892 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276452 | BC099838.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276452 | NM_178646.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276452 | AK036241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276452 | BC099838.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276452 | NM_178646.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276452 | AK036241.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93913 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109944 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276452 | BC099838.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276452 | NM_178646.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276452 | AK036241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276452 | BC099838.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276452 | NM_178646.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276452 | AK036241.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30344896 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109944 | - | - |
| Whole Mouse Genome G4122A | A_51_P276452 | BC099838.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276452 | NM_178646.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276452 | AK036241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276452 | BC099838.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276452 | NM_178646.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276452 | AK036241.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108745 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109944 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12156072 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344888 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30344892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30344881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30344930 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424745 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440377_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449646_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460743_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424743 | - | - |
| Mouse Genome 430 2.0 Array | 1460743_at | - | - |
| Mouse Genome 430 2.0 Array | 1449646_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1440377_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440377_at | - | - |
| Mouse Genome 430 2.0 Array | 1449646_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1460743_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105734_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105734_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500003897.mm.1 | NM_178646.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003897.mm.1 | NM_178646.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003897.mm.1 | TC1500000566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500003900.mm.1 | NM_178646.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003900.mm.1 | NM_178646.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003900.mm.1 | TC1500000566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500003899.mm.1 | NM_178646.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003899.mm.1 | NM_178646.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003899.mm.1 | TC1500000566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500003901.mm.1 | NM_178646.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003901.mm.1 | NM_178646.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003901.mm.1 | TC1500000566.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1449646_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460743_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460743_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449646_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440377_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440377_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 469386 | - | - |
| ABI Mouse Genome Survey Microarray | 583188 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 105360131 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 110451 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110451 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221043 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246540 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221043 | - | - |
| mouse-6 v1.1 (Illumina) | scl47812.1_441-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105360131 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110451 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47812.1_441-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110451 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105360131 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221043 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221043 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246540 | - | - |

Tigd5
Genomic Map