Assay Details
| Gene Symbol: Entpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032057.1 | - | 4 - 5 | 351 | 122 | |
| AK046218.1 | - | 4 - 5 | 340 | 122 | ||
| AK087561.1 | - | 4 - 5 | 338 | 122 | ||
| AY376710.1 | - | 4 - 5 | 332 | 122 | ||
| AY714060.1 | - | 4 - 5 | 335 | 122 | ||
| BC079871.1 | - | 4 - 5 | 335 | 6831925 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032057.1 transcripts.
Set Membership: |
|

Entpd3
Genomic Map