Assay Details
| Gene Symbol: Depdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178683.4 | NP_848798.1 | 8 - 9 | 1151 | 90 | |
| GenBank mRNA | AK036550.1 | - | 8 - 9 | 1110 | 90 | |
| AK043283.1 | - | 1108 | 90 | |||
| AK077676.1 | - | 8 - 9 | 1152 | 90 | ||
| AK083480.1 | - | 8 - 9 | 1147 | 90 | ||
| BC048246.1 | - | 1 - 2 | 183 | 90 | ||
| BC152330.1 | - | 8 - 9 | 1156 | 40141538 | 90 | |
| BC152331.1 | - | 8 - 9 | 1156 | 40141540 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Depdc1b
Genomic Map