Assay Details
| Gene Symbol: Eno4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178689.4 | NP_848804.2 | 11 - 12 | 1582 | 72 | |
| GenBank mRNA | AK032395.1 | - | 11 - 12 | 1585 | 72 | |
| AK076682.1 | - | 7 - 8 | 762 | 72 | ||
| BC023285.1 | - | 8 - 9 | 1025 | 3984456 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eno4
Genomic Map