Assay Details
| Gene Symbol: Eno4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178689.4 | NP_848804.2 | 7 - 8 | 1028 | 101 | |
| GenBank mRNA | AK032395.1 | - | 7 - 8 | 1031 | 101 | |
| AK076682.1 | - | 3 - 4 | 208 | 101 | ||
| BC023285.1 | - | 4 - 5 | 471 | 3984456 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eno4
Genomic Map