Assay Details
| Gene Symbol: Eno4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178689.4 | NP_848804.2 | 8 - 9 | 1087 | 96 | |
| GenBank mRNA | AK032395.1 | - | 8 - 9 | 1090 | 96 | |
| AK076682.1 | - | 4 - 5 | 267 | 96 | ||
| BC023285.1 | - | 5 - 6 | 530 | 3984456 | 96 | |
More Information
Set Membership: |
|

Eno4
Genomic Map