Assay Details
| Gene Symbol: Rab3gap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178690.4 | NP_848805.2 | 3 - 4 | 192 | 61 | |
| GenBank mRNA | AK076399.1 | - | 3 - 4 | 175 | 61 | |
| AK080432.1 | - | 3 - 4 | 174 | 61 | ||
| AK134604.1 | - | 3 - 4 | 174 | 61 | ||
| AK164523.1 | - | 3 - 4 | 173 | 61 | ||
| AK170768.1 | - | 3 - 4 | 164 | 61 | ||
| AK172881.1 | - | 3 - 4 | 179 | 61 | ||
| BC046297.1 | - | 3 - 4 | 158 | 6486170 | 61 | |
| CJ084350.1 | - | 3 - 4 | 193 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab3gap1
Genomic Map