Assay Details
Gene Symbol: Dpy19l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178704.3 | NP_848819.2 | 8 - 9 | 1096 | 71 | |
GenBank mRNA | AF538322.1 | - | 8 - 9 | 1080 | 71 | |
AK031345.1 | - | 8 - 9 | 1088 | 71 | ||
AK033395.1 | - | 8 - 9 | 1053 | 71 | ||
AK034217.1 | - | 8 - 9 | 1097 | 71 | ||
AK045758.1 | - | 8 - 9 | 997 | 71 | ||
BC050065.1 | - | 8 - 9 | 1040 | 30016211 | 71 | |
BC059897.1 | - | 8 - 9 | 993 | 6821433 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
