| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600014739.mm.1 |
NM_001145977.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014739.mm.1 |
NM_001145977.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014739.mm.1 |
NM_178721.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014739.mm.1 |
NM_178721.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014739.mm.1 |
TC1600001849.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600014737.mm.1 |
NM_001145977.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014737.mm.1 |
NM_001145977.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014737.mm.1 |
NM_178721.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014737.mm.1 |
NM_178721.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014737.mm.1 |
TC1600001849.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600014738.mm.1 |
NM_001145977.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014738.mm.1 |
NM_001145977.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014738.mm.1 |
NM_178721.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014738.mm.1 |
NM_178721.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014738.mm.1 |
TC1600001849.mm.1 |
MTA TC |