Assay Details
| Gene Symbol: Ppm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178726.3 | NP_848841.2 | 2 - 3 | 1223 | 83 | |
| GenBank mRNA | AF117832.1 | - | 1 - 2 | 178 | 83 | |
| AK028275.1 | - | 2 - 3 | 265 | 83 | ||
| AK032529.1 | - | 2 - 3 | 1281 | 83 | ||
| AK035912.1 | - | 2 - 3 | 1226 | 83 | ||
| AK045724.1 | - | 2 - 3 | 1264 | 83 | ||
| AK131646.1 | - | 2 - 3 | 1210 | 83 | ||
| AK142789.1 | - | 2 - 3 | 1241 | 83 | ||
| AK147389.1 | - | 2 - 3 | 1224 | 83 | ||
| AK147876.1 | - | 2 - 3 | 1260 | 83 | ||
| AK165160.1 | - | 2 - 3 | 1256 | 83 | ||
| AK220523.1 | - | 2 - 3 | 1228 | 83 | ||
| AY184801.1 | - | 2 - 3 | 811 | 83 | ||
| BC096031.1 | - | 2 - 3 | 1223 | 6837066 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1l
Genomic Map