Assay Details
| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159963.1 | NP_001153435.1 | 3 - 4 | 489 | 140 | |
| NM_178729.4 | NP_848844.1 | 3 - 4 | 489 | 140 | ||
| GenBank mRNA | AK029957.1 | - | 3 - 4 | 437 | 140 | |
| AK041497.1 | - | 3 - 4 | 461 | 140 | ||
| AK054009.1 | - | 3 - 4 | 459 | 140 | ||
| AK082931.1 | - | 3 - 4 | 459 | 140 | ||
| AK085100.1 | - | 3 - 4 | 462 | 140 | ||
| AK088067.1 | - | 3 - 4 | 140 | |||
| AK150664.1 | - | 3 - 4 | 449 | 140 | ||
| AK159744.1 | - | 3 - 4 | 478 | 140 | ||
| AK171361.1 | - | 3 - 4 | 443 | 140 | ||
| AK171783.1 | - | 3 - 4 | 349 | 140 | ||
| BC023291.1 | - | 1 - 2 | 94 | 140 | ||
| BC047214.1 | - | 402 | 6466511 | 140 | ||
| BC113798.1 | - | 3 - 4 | 407 | 40062506 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl5
Genomic Map