Assay Details
| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159963.1 | NP_001153435.1 | 4 - 5 | 685 | 81 | |
| NM_178729.4 | NP_848844.1 | 4 - 5 | 685 | 81 | ||
| GenBank mRNA | AK029957.1 | - | 4 - 5 | 633 | 81 | |
| AK041497.1 | - | 4 - 5 | 657 | 81 | ||
| AK054009.1 | - | 4 - 5 | 655 | 81 | ||
| AK082931.1 | - | 4 - 5 | 655 | 81 | ||
| AK085100.1 | - | 4 - 5 | 658 | 81 | ||
| AK088067.1 | - | 4 - 5 | 652 | 81 | ||
| AK150664.1 | - | 4 - 5 | 645 | 81 | ||
| AK159744.1 | - | 4 - 5 | 674 | 81 | ||
| AK171361.1 | - | 4 - 5 | 639 | 81 | ||
| AK171783.1 | - | 4 - 5 | 545 | 81 | ||
| BC023291.1 | - | 2 - 3 | 290 | 81 | ||
| BC047214.1 | - | 598 | 6466511 | 81 | ||
| BC113798.1 | - | 4 - 5 | 603 | 40062506 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl5
Genomic Map