Assay Details
| Gene Symbol: Rasal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178785.3 | NP_848900.2 | 5 - 6 | 876 | 74 | |
| GenBank mRNA | AK030797.1 | - | 5 - 6 | 879 | 74 | |
| AK040573.1 | - | 5 - 6 | 864 | 74 | ||
| AK041479.1 | - | 6 - 7 | 734 | 74 | ||
| AK088449.1 | - | 5 - 6 | 838 | 74 | ||
| AK088987.1 | - | 5 - 6 | 854 | 74 | ||
| AK131128.1 | - | 7 - 8 | 783 | 74 | ||
| AK178305.1 | - | 2 - 3 | 108 | 74 | ||
| BC132341.1 | - | 7 - 8 | 782 | 40130618 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasal3
Genomic Map