Assay Details
| Gene Symbol: Rasal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178785.3 | NP_848900.2 | 6 - 7 | 1002 | 86 | |
| GenBank mRNA | AK030797.1 | - | 6 - 7 | 1005 | 86 | |
| AK040573.1 | - | 6 - 7 | 990 | 86 | ||
| AK041479.1 | - | 7 - 8 | 860 | 86 | ||
| AK088449.1 | - | 6 - 7 | 964 | 86 | ||
| AK088987.1 | - | 6 - 7 | 980 | 86 | ||
| AK131128.1 | - | 8 - 9 | 909 | 86 | ||
| BC132341.1 | - | 8 - 9 | 908 | 40130618 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasal3
Genomic Map