Assay Details
| Gene Symbol: Ccbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178793.4 | NP_848908.1 | 3 - 4 | 345 | 94 | |
| GenBank mRNA | AK028377.1 | - | 3 - 4 | 343 | 94 | |
| AK035153.1 | - | 3 - 4 | 340 | 94 | ||
| AK039742.1 | - | 3 - 4 | 275 | 94 | ||
| AK220435.1 | - | 3 - 4 | 315 | 94 | ||
| BC103803.1 | - | 3 - 4 | 295 | 30088062 | 94 | |
| BC152322.1 | - | 3 - 4 | 314 | 40140631 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbe1
Genomic Map