Assay Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178880.4 | NP_849211.3 | 9 - 10 | 7174 | 118 | |
| GenBank mRNA | AK035150.1 | - | 2 - 3 | 2665 | 118 | |
| AK122418.1 | - | 8 - 9 | 4290 | 118 | ||
| BC145121.1 | - | 9 - 10 | 7114 | 116 | ||
| CB849818.2 | - | 2 - 3 | 272 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Son
Genomic Map