Assay Details
| Gene Symbol: D2hgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178882.3 | NP_849213.2 | 6 - 7 | 999 | 67 | |
| GenBank mRNA | AK088200.1 | - | 946 | 67 | ||
| AK088554.1 | - | 946 | 67 | |||
| AK141691.1 | - | 6 - 7 | 1229 | 67 | ||
| AK170226.1 | - | 1000 | 67 | |||
| BC023277.1 | - | 4006813 | 67 | |||
| BC117794.2 | - | 6 - 7 | 937 | 40092173 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

D2hgdh
Genomic Map