Assay Details
| Gene Symbol: Coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164804.1 | NP_001158276.1 | 3 - 4 | 427 | 80 | |
| NM_178893.4 | NP_849224.2 | 4 - 5 | 652 | 80 | ||
| GenBank mRNA | AK030097.1 | - | 4 - 5 | 655 | 80 | |
| AK030971.1 | - | 3 - 4 | 384 | 80 | ||
| AK044060.1 | - | 3 - 4 | 389 | 80 | ||
| AK054394.1 | - | 3 - 4 | 428 | 80 | ||
| AK152490.1 | - | 3 - 4 | 416 | 80 | ||
| AK153134.1 | - | 3 - 4 | 416 | 80 | ||
| AK154247.1 | - | 3 - 4 | 384 | 80 | ||
| AK170184.1 | - | 3 - 4 | 385 | 80 | ||
| BC031164.1 | - | 1 - 2 | 74 | 80 | ||
| BC138731.1 | - | 3 - 4 | 452 | 8861753 | 80 | |
| BC138732.1 | - | 3 - 4 | 452 | 8861754 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coro2a
Genomic Map