Assay Details
| Gene Symbol: Coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164804.1 | NP_001158276.1 | 5 - 6 | 767 | 96 | |
| NM_178893.4 | NP_849224.2 | 6 - 7 | 992 | 96 | ||
| GenBank mRNA | AK030097.1 | - | 6 - 7 | 995 | 96 | |
| AK030971.1 | - | 5 - 6 | 724 | 96 | ||
| AK044060.1 | - | 5 - 6 | 729 | 96 | ||
| AK054394.1 | - | 5 - 6 | 768 | 96 | ||
| AK152490.1 | - | 5 - 6 | 756 | 96 | ||
| AK153134.1 | - | 5 - 6 | 756 | 96 | ||
| AK154247.1 | - | 5 - 6 | 724 | 96 | ||
| AK170184.1 | - | 5 - 6 | 725 | 96 | ||
| BC031164.1 | - | 3 - 4 | 414 | 96 | ||
| BC138731.1 | - | 5 - 6 | 792 | 8861753 | 96 | |
| BC138732.1 | - | 5 - 6 | 792 | 8861754 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coro2a
Genomic Map