Assay Details
| Gene Symbol: Gars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180678.3 | NP_851009.2 | 14 - 15 | 1815 | 96 | |
| GenBank mRNA | AK012758.2 | - | 14 - 15 | 1810 | 96 | |
| AK165857.1 | - | 14 - 15 | 1763 | 96 | ||
| BC021747.1 | - | 14 - 15 | 1796 | 4192628 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gars
Genomic Map